Product Details

SNP ID
rs148200469
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:62707586 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAAATATCCTGACATGGTATCTG[G/T]GGGGGTACTTGGAGTGGCATTTGGG
Phenotype
MIM: 605716
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KCNH5 PubMed Links

Gene Details

Gene
KCNH5
Gene Name
potassium voltage-gated channel subfamily H member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139318.4 2744 Silent Mutation NP_647479.2
NM_172375.2 2744 UTR 3 NP_758963.1

View Full Product Details