Product Details

SNP ID
rs149824477
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:60249460 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGCTCTCTGCCTCCCTCTCCAAC[A/G]CCCGGATGATCTGAGCCGCGAGGGC
Phenotype
MIM: 606108
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101927702 PubMed Links

Gene Details

Gene
LOC101927702
Gene Name
uncharacterized LOC101927702
There are no transcripts associated with this gene.

Gene
PPM1A
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021003.4 216 UTR 5 NP_066283.1
NM_177951.2 216 UTR 5 NP_808820.1
NM_177952.2 216 Intron NP_808821.2
XM_005267779.1 216 Intron XP_005267836.1
XM_005267781.1 216 UTR 5 XP_005267838.1
XM_011536872.1 216 UTR 5 XP_011535174.1
XM_011536876.1 216 UTR 5 XP_011535178.1
XM_011536877.1 216 UTR 5 XP_011535179.1
XM_011536878.1 216 UTR 5 XP_011535180.1
XM_011536879.2 216 Intron XP_011535181.1
XM_011536880.2 216 Intron XP_011535182.1
XM_011536881.2 216 Intron XP_011535183.1
XM_011536882.2 216 Intron XP_011535184.1
XM_011536883.1 216 UTR 5 XP_011535185.1
XM_011536884.1 216 Intron XP_011535186.1
XM_017021381.1 216 UTR 5 XP_016876870.1
XM_017021382.1 216 UTR 5 XP_016876871.1
XM_017021383.1 216 UTR 5 XP_016876872.1
XM_017021384.1 216 Intron XP_016876873.1
XM_017021385.1 216 UTR 5 XP_016876874.1
XM_017021386.1 216 Intron XP_016876875.1
XM_017021387.1 216 UTR 5 XP_016876876.1

View Full Product Details