Product Details

SNP ID
rs149988341
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:32092345 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCCACTAAAGAAACATGTCTTA[A/G]TGGCCAAAATTGTACAGACATTAAA
Phenotype
MIM: 602680
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP5 PubMed Links

Gene Details

Gene
ARHGAP5
Gene Name
Rho GTPase activating protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001030055.1 1960 Missense Mutation AAT,AGT N559S NP_001025226.1
NM_001173.2 1960 Missense Mutation AAT,AGT N559S NP_001164.2
XM_005267635.3 1960 Missense Mutation AAT,AGT N559S XP_005267692.1
XM_005267636.3 1960 Missense Mutation AAT,AGT N559S XP_005267693.1
XM_017021287.1 1960 Missense Mutation AAT,AGT N559S XP_016876776.1
XM_017021288.1 1960 Missense Mutation AAT,AGT N559S XP_016876777.1

View Full Product Details