Product Details

SNP ID
rs150248677
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24148849 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCTATTGAGAGAGATTGCTCCT[A/G]GCCCCCTCACCACACCCTCTGTCCC
Phenotype
MIM: 600654 MIM: 602161 MIM: 612487
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EMC9 PubMed Links

Gene Details

Gene
EMC9
Gene Name
ER membrane protein complex subunit 9
There are no transcripts associated with this gene.

Gene
MIR7703
Gene Name
microRNA 7703
There are no transcripts associated with this gene.

Gene
PSME1
Gene Name
proteasome activator subunit 1
There are no transcripts associated with this gene.

Gene
PSME2
Gene Name
proteasome activator subunit 2
There are no transcripts associated with this gene.

Gene
RNF31
Gene Name
ring finger protein 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001310332.1 509 Missense Mutation AGC,GGC S51G NP_001297261.1
NM_017999.4 509 Missense Mutation AGC,GGC S202G NP_060469.4

View Full Product Details