Product Details

SNP ID
rs150356603
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20456834 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGTGGGCCTGCTTTCCCGCCAGT[A/G]CCCACTCAAAGTTTCTTACGGCATA
Phenotype
MIM: 107748 MIM: 610107 MIM: 609865
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APEX1 PubMed Links

Gene Details

Gene
APEX1
Gene Name
apurinic/apyrimidinic endodeoxyribonuclease 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244249.1 679 Missense Mutation TAC,TGC Y138C NP_001231178.1
NM_001641.3 679 Missense Mutation TAC,TGC Y138C NP_001632.2
NM_080648.2 679 Missense Mutation TAC,TGC Y138C NP_542379.1
NM_080649.2 679 Missense Mutation TAC,TGC Y138C NP_542380.1
Gene
OSGEP
Gene Name
O-sialoglycoprotein endopeptidase
There are no transcripts associated with this gene.

Gene
TMEM55B
Gene Name
transmembrane protein 55B
There are no transcripts associated with this gene.

View Full Product Details