Product Details

SNP ID
rs150391208
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:21468681 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGATCTGGCTCACCTTGTAATGTC[A/G]TACACCAGCAGTGCTCCAGCTGCTC
Phenotype
MIM: 607466 MIM: 614032
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAB2B PubMed Links

Gene Details

Gene
RAB2B
Gene Name
RAB2B, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163380.1 286 Silent Mutation TAC,TAT Y40Y NP_001156852.1
NM_032846.3 286 Silent Mutation TAC,TAT Y86Y NP_116235.2
XM_017021710.1 286 Silent Mutation TAC,TAT Y41Y XP_016877199.1
XM_017021711.1 286 Silent Mutation TAC,TAT Y21Y XP_016877200.1
XM_017021712.1 286 Silent Mutation TAC,TAT Y17Y XP_016877201.1
Gene
TOX4
Gene Name
TOX high mobility group box family member 4
There are no transcripts associated with this gene.

View Full Product Details