Product Details

SNP ID
rs150417989
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:19876283 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCAAGACAATGGATGTGGGCAAT[A/G]AGTCTACCATGTCTGAATTTGTTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR4K2 PubMed Links

Gene Details

Gene
OR4K2
Gene Name
olfactory receptor family 4 subfamily K member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005501.1 16 Missense Mutation AAG,GAG K6E NP_001005501.1

View Full Product Details