Product Details

SNP ID
rs150616937
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:77406774 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAAGAATCGTAGGATCTTGGAGA[G/T]CAGCTATGACTCCCTTATTGGCCCC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NOXRED1 PubMed Links

Gene Details

Gene
NOXRED1
Gene Name
NADP dependent oxidoreductase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113475.2 712 Missense Mutation GAT,GCT D211A NP_001106946.1
XM_005267330.3 712 Missense Mutation GAT,GCT D211A XP_005267387.1
XM_011536427.2 712 Missense Mutation GAT,GCT D211A XP_011534729.1
XM_011536428.2 712 Missense Mutation GAT,GCT D211A XP_011534730.1
XM_011536429.2 712 Missense Mutation GAT,GCT D121A XP_011534731.1
XM_017020969.1 712 Intron XP_016876458.1
XM_017020970.1 712 Intron XP_016876459.1

View Full Product Details