Product Details

SNP ID
rs150768256
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:52643570 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGGCACATATCAGCTTGTTTTCT[C/T]TAATGAATATTCTGTAACAAGTTCC
Phenotype
MIM: 615435
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ERO1A PubMed Links

Gene Details

Gene
ERO1A
Gene Name
endoplasmic reticulum oxidoreductase 1 alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014584.2 1664 Silent Mutation TAA,TAG *469* NP_055399.1
XM_011536684.2 1664 Silent Mutation TAA,TAG *465* XP_011534986.1
Gene
GPR137C
Gene Name
G protein-coupled receptor 137C
There are no transcripts associated with this gene.

View Full Product Details