Product Details

SNP ID
rs150820756
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:94442621 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCACCATGGTTACCCTGCAACTGG[G/T]TTGACAACTTTTCCCAGGAAGAGTA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SERPINA11 PubMed Links

Gene Details

Gene
SERPINA11
Gene Name
serpin family A member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080451.1 869 Missense Mutation AAA,AAC K418N NP_001073920.1
XM_006720105.3 869 Missense Mutation AAA,AAC K214N XP_006720168.1

View Full Product Details