Product Details

SNP ID
rs150934075
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20455917 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATATATTACTCATTTTATAGAGC[C/T]AGAGGCCAAGAAGAGTAAGACGGCC
Phenotype
MIM: 107748 MIM: 610107 MIM: 609865
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APEX1 PubMed Links

Gene Details

Gene
APEX1
Gene Name
apurinic/apyrimidinic endodeoxyribonuclease 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244249.1 328 Missense Mutation CCA,CTA P21L NP_001231178.1
NM_001641.3 328 Missense Mutation CCA,CTA P21L NP_001632.2
NM_080648.2 328 Missense Mutation CCA,CTA P21L NP_542379.1
NM_080649.2 328 Missense Mutation CCA,CTA P21L NP_542380.1
Gene
OSGEP
Gene Name
O-sialoglycoprotein endopeptidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017807.3 328 Intron NP_060277.1
Gene
TMEM55B
Gene Name
transmembrane protein 55B
There are no transcripts associated with this gene.

View Full Product Details