Product Details
- SNP ID
-
rs111499497
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:34088080 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCTCATGTCAGGATCACTTGTGTT[A/G]ACCACTTTAGGCAGAAGCACAAATA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EMC7
PubMed Links
Gene Details
- Gene
- EMC7
- Gene Name
- ER membrane protein complex subunit 7
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020154.2 |
562 |
Silent Mutation |
GTC,GTT |
V183V |
NP_064539.1 |
View Full Product Details