Product Details

SNP ID
rs111553256
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:42211744 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACCTTTCAAAGTGTGTGATCAT[C/T]CGTTCCTAGGGAAAAAAAAAAAGGT
Phenotype
MIM: 612188
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM87A PubMed Links

Gene Details

Gene
TMEM87A
Gene Name
transmembrane protein 87A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001110503.2 1677 Intron NP_001103973.1
NM_001286487.1 1677 Silent Mutation CGA,CGG R483R NP_001273416.1
NM_015497.4 1677 Silent Mutation CGA,CGG R544R NP_056312.2
XM_005254287.1 1677 Silent Mutation CGA,CGG R545R XP_005254344.1
XM_006720468.2 1677 Silent Mutation CGA,CGG R404R XP_006720531.1
XM_011521443.2 1677 Silent Mutation CGA,CGG R484R XP_011519745.1
XM_011521444.2 1677 Intron XP_011519746.1
XM_017022067.1 1677 Silent Mutation CGA,CGG R403R XP_016877556.1
Gene
VPS39
Gene Name
VPS39, HOPS complex subunit
There are no transcripts associated with this gene.

View Full Product Details