Product Details

SNP ID
rs113183884
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:42160763 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCAGGATGCTGGGCTCAAGTGTC[A/G]GCTGGGTTTACCTCTTTGGAACAGA
Phenotype
MIM: 612188
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLA2G4F PubMed Links

Gene Details

Gene
PLA2G4F
Gene Name
phospholipase A2 group IVF
There are no transcripts associated with this gene.

Gene
VPS39
Gene Name
VPS39, HOPS complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301138.1 2791 Silent Mutation GCC,GCT A884A NP_001288067.1
NM_015289.3 2791 Silent Mutation GCC,GCT A873A NP_056104.2
XM_017022036.1 2791 Intron XP_016877525.1
XM_017022037.1 2791 Intron XP_016877526.1
XM_017022038.1 2791 Silent Mutation GCC,GCT A749A XP_016877527.1
XM_017022039.1 2791 Silent Mutation GCC,GCT A516A XP_016877528.1

View Full Product Details