Product Details

SNP ID
rs113970475
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:99715706 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCATTGCAAGAAAATTTTATCCC[C/T]CCAGAAGTATTTTATTACTAAAGAA
Phenotype
MIM: 600660
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LYSMD4 PubMed Links

Gene Details

Gene
LYSMD4
Gene Name
LysM domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284417.1 5214 Intron NP_001271346.1
NM_001284418.1 5214 Intron NP_001271347.1
NM_001284419.1 5214 Intron NP_001271348.1
NM_001284420.1 5214 Intron NP_001271349.1
NM_001284421.1 5214 Intron NP_001271350.1
NM_001284422.1 5214 Intron NP_001271351.1
NM_152449.3 5214 Intron NP_689662.2
XM_006720397.3 5214 Intron XP_006720460.2
XM_011521241.1 5214 Intron XP_011519543.1
XM_011521242.2 5214 UTR 3 XP_011519544.1
XM_011521243.2 5214 Intron XP_011519545.1
XM_011521244.1 5214 Intron XP_011519546.1
XM_011521245.2 5214 UTR 3 XP_011519547.1
XM_017021926.1 5214 Intron XP_016877415.1
XM_017021927.1 5214 Intron XP_016877416.1
XM_017021928.1 5214 Intron XP_016877417.1
XM_017021929.1 5214 Intron XP_016877418.1
XM_017021930.1 5214 Intron XP_016877419.1
Gene
MEF2A
Gene Name
myocyte enhancer factor 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130926.2 5214 UTR 3 NP_001124398.1
NM_001130927.2 5214 UTR 3 NP_001124399.1
NM_001130928.2 5214 UTR 3 NP_001124400.1
NM_001171894.2 5214 UTR 3 NP_001165365.1
NM_001319206.1 5214 UTR 3 NP_001306135.1
NM_005587.3 5214 UTR 3 NP_005578.2
XM_005254914.2 5214 UTR 3 XP_005254971.1
XM_005254915.2 5214 UTR 3 XP_005254972.1
XM_005254916.3 5214 Intron XP_005254973.1
XM_011521571.2 5214 UTR 3 XP_011519873.1
XM_011521572.2 5214 UTR 3 XP_011519874.1
XM_011521573.2 5214 UTR 3 XP_011519875.1
XM_011521576.2 5214 UTR 3 XP_011519878.1
XM_011521577.2 5214 UTR 3 XP_011519879.1
XM_011521578.2 5214 UTR 3 XP_011519880.1
XM_011521579.2 5214 UTR 3 XP_011519881.1
XM_011521581.2 5214 UTR 3 XP_011519883.1
XM_011521582.2 5214 UTR 3 XP_011519884.1
XM_011521583.2 5214 UTR 3 XP_011519885.1
XM_011521585.2 5214 UTR 3 XP_011519887.1
XM_011521586.2 5214 UTR 3 XP_011519888.1
XM_011521587.2 5214 UTR 3 XP_011519889.1
XM_011521590.2 5214 Intron XP_011519892.1
XM_017022190.1 5214 UTR 3 XP_016877679.1
XM_017022191.1 5214 UTR 3 XP_016877680.1
XM_017022192.1 5214 UTR 3 XP_016877681.1
XM_017022193.1 5214 UTR 3 XP_016877682.1
XM_017022194.1 5214 UTR 3 XP_016877683.1
XM_017022195.1 5214 UTR 3 XP_016877684.1
XM_017022196.1 5214 UTR 3 XP_016877685.1
XM_017022197.1 5214 UTR 3 XP_016877686.1
XM_017022198.1 5214 UTR 3 XP_016877687.1
XM_017022199.1 5214 UTR 3 XP_016877688.1
XM_017022200.1 5214 UTR 3 XP_016877689.1
XM_017022201.1 5214 UTR 3 XP_016877690.1
XM_017022202.1 5214 UTR 3 XP_016877691.1
XM_017022203.1 5214 Intron XP_016877692.1
XM_017022204.1 5214 Intron XP_016877693.1

View Full Product Details