Product Details

SNP ID
rs114347421
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40355331 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGATTTCTTGGGTCTGGCATCC[A/G]CTCAGGAACAGGTATGAGCTCGCAC
Phenotype
MIM: 607503
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DISP2 PubMed Links

Gene Details

Gene
DISP2
Gene Name
dispatched RND transporter family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033510.2 973 Intron NP_277045.1
XM_011522125.2 973 UTR 5 XP_011520427.1
XM_017022687.1 973 UTR 5 XP_016878176.1
Gene
PHGR1
Gene Name
proline, histidine and glycine rich 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145643.1 973 Intron NP_001139115.1

View Full Product Details