Product Details

SNP ID
rs114810334
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:80951248 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGTTGAATGTCTTTGGCTCAGTT[C/G]ATTTAAAAAAGATATCTATTTGAAA
Phenotype
MIM: 608366 MIM: 607783
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CEMIP PubMed Links

Gene Details

Gene
CEMIP
Gene Name
cell migration inducing hyaluronan binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001293298.1 6698 UTR 3 NP_001280227.1
NM_001293304.1 6698 UTR 3 NP_001280233.1
NM_018689.2 6698 UTR 3 NP_061159.1
Gene
MESDC2
Gene Name
mesoderm development candidate 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015154.2 6698 Intron NP_055969.1

View Full Product Details