Product Details

SNP ID
rs115424730
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:45402623 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCTCTGCACTGCCTGGCCTCGGC[A/G]GGACGGAGCGGACGGCTTTGTGCAG
Phenotype
MIM: 602360
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GATM PubMed Links

Gene Details

Gene
GATM
Gene Name
glycine amidinotransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321015.1 303 Intron NP_001307944.1
NM_001482.2 303 Intron NP_001473.1
Gene
SPATA5L1
Gene Name
spermatogenesis associated 5 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323640.1 303 Missense Mutation CAG,CGG Q65R NP_001310569.1
NM_024063.2 303 Missense Mutation CAG,CGG Q65R NP_076968.2

View Full Product Details