Product Details

SNP ID
rs115550738
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:87863538 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCCAAAAGTCTTTTTTTTTTTTT[G/T]TATGTGTGTCATTTCAAACAAAAAT
Phenotype
MIM: 191316
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NTRK3 PubMed Links

Gene Details

Gene
NTRK3
Gene Name
neurotrophic receptor tyrosine kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007156.2 15842 Intron NP_001007157.1
NM_001012338.2 15842 Intron NP_001012338.1
NM_001243101.1 15842 Intron NP_001230030.1
NM_001320134.1 15842 Intron NP_001307063.1
NM_001320135.1 15842 Intron NP_001307064.1
NM_002530.3 15842 Intron NP_002521.2
XM_006720543.3 15842 UTR 3 XP_006720606.1
XM_006720544.3 15842 UTR 3 XP_006720607.1
XM_006720545.3 15842 Intron XP_006720608.1
XM_006720548.3 15842 Intron XP_006720611.1
XM_006720549.3 15842 Intron XP_006720612.1
XM_006720550.3 15842 Intron XP_006720613.1
XM_011521637.2 15842 Intron XP_011519939.1
XM_011521638.2 15842 Intron XP_011519940.1
XM_017022240.1 15842 UTR 3 XP_016877729.1
XM_017022241.1 15842 UTR 3 XP_016877730.1
XM_017022242.1 15842 Intron XP_016877731.1
XM_017022243.1 15842 UTR 3 XP_016877732.1
XM_017022244.1 15842 UTR 3 XP_016877733.1
XM_017022245.1 15842 UTR 3 XP_016877734.1
XM_017022246.1 15842 UTR 3 XP_016877735.1
XM_017022247.1 15842 Intron XP_016877736.1
XM_017022248.1 15842 Intron XP_016877737.1
XM_017022249.1 15842 Intron XP_016877738.1
XM_017022250.1 15842 Intron XP_016877739.1
XM_017022251.1 15842 UTR 3 XP_016877740.1
XM_017022252.1 15842 UTR 3 XP_016877741.1
XM_017022253.1 15842 UTR 3 XP_016877742.1
XM_017022254.1 15842 UTR 3 XP_016877743.1

View Full Product Details