Product Details
- SNP ID
-
rs115629509
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:43278167 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAGCTGCCCAGTTCTCTATTGGGG[C/T]GGGGAGGGGCAGGCTGTTGGAGAAG
- Phenotype
-
MIM: 606776
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TGM7
PubMed Links
Gene Details
- Gene
- TGM7
- Gene Name
- transglutaminase 7
View Full Product Details