Product Details

SNP ID
rs115780816
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:68801868 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATAAACTCTGAGGAATGGGGACA[A/G]GAACAAGGCCTGACTCCTCTGGGGA
Phenotype
MIM: 600832
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANP32A PubMed Links

Gene Details

Gene
ANP32A
Gene Name
acidic nuclear phosphoprotein 32 family member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006305.3 Intron NP_006296.1
Gene
ANP32A-IT1
Gene Name
ANP32A intronic transcript 1
There are no transcripts associated with this gene.

Gene
MIR4312
Gene Name
microRNA 4312
There are no transcripts associated with this gene.

View Full Product Details