Product Details

SNP ID
rs116525439
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:68784470 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCGAGTCAGGGGCCTCCTTGTC[A/G]TCCCGGTCATAGCCGTCGAGATATG
Phenotype
MIM: 600832
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANP32A PubMed Links

Gene Details

Gene
ANP32A
Gene Name
acidic nuclear phosphoprotein 32 family member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006305.3 Intron NP_006296.1

View Full Product Details