Product Details

SNP ID
rs117108060
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:68191355 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACAATGTCAGTTTGTATAAAAGT[A/G]CCAAGTGAAAATATTTATTACATGC
Phenotype
MIM: 603566
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CALML4 PubMed Links

Gene Details

Gene
CALML4
Gene Name
calmodulin like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031733.2 6223 UTR 3 NP_001026903.2
NM_001286694.1 6223 UTR 3 NP_001273623.1
NM_001286695.1 6223 UTR 3 NP_001273624.1
NM_033429.2 6223 UTR 3 NP_219501.2
Gene
PIAS1
Gene Name
protein inhibitor of activated STAT 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320687.1 6223 UTR 3 NP_001307616.1
NM_016166.2 6223 UTR 3 NP_057250.1
XM_011522126.2 6223 Intron XP_011520428.1
XM_011522127.2 6223 Intron XP_011520429.1
XM_017022688.1 6223 Intron XP_016878177.1
XM_017022689.1 6223 Intron XP_016878178.1
XM_017022690.1 6223 Intron XP_016878179.1
XM_017022691.1 6223 Intron XP_016878180.1

View Full Product Details