Product Details

SNP ID
rs117112941
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:81312886 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGATGATTCTCTATGACAGTCAG[A/G]GAAACGTGGTCTCTGAGAGTGATGG
Phenotype
MIM: 603035 MIM: 607050
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL16 PubMed Links

Gene Details

Gene
IL16
Gene Name
interleukin 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172128.1 1611 Intron NP_001165599.1
NM_004513.5 1611 Intron NP_004504.3
NM_172217.3 1611 Intron NP_757366.2
XM_005254342.3 1611 Intron XP_005254399.1
XM_005254346.4 1611 Intron XP_005254403.1
XM_011521519.2 1611 Intron XP_011519821.1
XM_011521520.2 1611 Intron XP_011519822.1
XM_017022140.1 1611 Intron XP_016877629.1
XM_017022141.1 1611 Intron XP_016877630.1
XM_017022142.1 1611 Intron XP_016877631.1
XM_017022143.1 1611 Intron XP_016877632.1
XM_017022144.1 1611 Intron XP_016877633.1
XM_017022145.1 1611 Intron XP_016877634.1
XM_017022146.1 1611 Intron XP_016877635.1
Gene
STARD5
Gene Name
StAR related lipid transfer domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181900.2 1611 UTR 3 NP_871629.1
XM_006720703.1 1611 Intron XP_006720766.1
XM_017022639.1 1611 UTR 3 XP_016878128.1

View Full Product Details