Product Details

SNP ID
rs117118484
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:66388402 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTTTTGGTTGTTGTTTGCCCAA[C/T]CTTCAATAAGGATTGGGTTCTTAAT
Phenotype
MIM: 176872 MIM: 610716
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAP2K1 PubMed Links

Gene Details

Gene
MAP2K1
Gene Name
mitogen-activated protein kinase kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002755.3 Intron NP_002746.1
XM_011521783.2 Intron XP_011520085.1
XM_017022411.1 Intron XP_016877900.1
XM_017022412.1 Intron XP_016877901.1
XM_017022413.1 Intron XP_016877902.1
Gene
TIPIN
Gene Name
TIMELESS interacting protein
There are no transcripts associated with this gene.

View Full Product Details