Product Details

SNP ID
rs138702546
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:82759339 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATGGAAGACAGGAAACTGGACAT[C/T]AGAATGTTCTAATAGAAAGTGACAT
Phenotype
MIM: 612675
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FSD2 PubMed Links

Gene Details

Gene
FSD2
Gene Name
fibronectin type III and SPRY domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007122.3 2442 UTR 3 NP_001007123.1
NM_001281805.1 2442 UTR 3 NP_001268734.1
NM_001281806.1 2442 UTR 3 NP_001268735.1
XM_005272425.4 2442 UTR 3 XP_005272482.1
XM_011521235.2 2442 UTR 3 XP_011519537.1
XM_017021924.1 2442 Intron XP_016877413.1
Gene
SCARNA15
Gene Name
small Cajal body-specific RNA 15
There are no transcripts associated with this gene.

Gene
SNHG21
Gene Name
small nucleolar RNA host gene 21
There are no transcripts associated with this gene.

View Full Product Details