Product Details

SNP ID
rs138839022
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:39252532 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGTCTCCTGGGTGGAAGAACATG[A/T]GTTCAACAAAGCGTGTGTGCACTAT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C15orf54 PubMed Links

Gene Details

Gene
C15orf54
Gene Name
chromosome 15 open reading frame 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302797.1 669 Missense Mutation AGT,TGT S133C NP_001289726.1
NM_207445.3 669 Missense Mutation AGT,TGT S133C NP_997328.1

View Full Product Details