Product Details

SNP ID
rs138868218
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:75649571 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGCCTACCCCAGCCAGCACATG[C/G]CCTTCCGGCCCAAGCCACCACTGGA
Phenotype
MIM: 612980
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IMP3 PubMed Links

Gene Details

Gene
IMP3
Gene Name
IMP3, U3 small nucleolar ribonucleoprotein
There are no transcripts associated with this gene.

Gene
LOC107984799
Gene Name
translation initiation factor IF-2-like
There are no transcripts associated with this gene.

Gene
SNX33
Gene Name
sorting nexin 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318146.1 565 Missense Mutation CCC,GCC P157A NP_001305075.1
NM_153271.1 565 Missense Mutation CCC,GCC P157A NP_695003.1

View Full Product Details