Product Details

SNP ID
rs139444854
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:78341176 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACCACTGAGATCAACTTCAAGGT[C/T]GGAGAAGGCTTTGAGGAGGAGACCG
Phenotype
MIM: 180230
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRABP1 PubMed Links

Gene Details

Gene
CRABP1
Gene Name
cellular retinoic acid binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004378.2 309 Silent Mutation GTC,GTT V68V NP_004369.1

View Full Product Details