Product Details
- SNP ID
-
rs139473906
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:70052486 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGTTGCTGCTCTCCATGCCCACAGC[C/T]AGCCACTCCCCAGTGGGGCAGTAGC
- Phenotype
-
MIM: 600190
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TLE3
PubMed Links
Gene Details
- Gene
- TLE3
- Gene Name
- transducin like enhancer of split 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001105192.2 |
3165 |
Silent Mutation |
CTA,CTG |
L671L |
NP_001098662.1 |
NM_001282979.1 |
3165 |
Silent Mutation |
CTA,CTG |
L666L |
NP_001269908.1 |
NM_001282980.1 |
3165 |
Silent Mutation |
CTA,CTG |
L669L |
NP_001269909.1 |
NM_001282981.1 |
3165 |
Silent Mutation |
CTA,CTG |
L664L |
NP_001269910.1 |
NM_001282982.1 |
3165 |
Silent Mutation |
CTA,CTG |
L601L |
NP_001269911.1 |
NM_005078.3 |
3165 |
Silent Mutation |
CTA,CTG |
L674L |
NP_005069.2 |
NM_020908.2 |
3165 |
Silent Mutation |
CTA,CTG |
L662L |
NP_065959.1 |
XM_005254622.4 |
3165 |
Silent Mutation |
CTA,CTG |
L681L |
XP_005254679.1 |
XM_005254623.3 |
3165 |
Silent Mutation |
CTA,CTG |
L676L |
XP_005254680.1 |
XM_005254625.3 |
3165 |
Silent Mutation |
CTA,CTG |
L672L |
XP_005254682.1 |
XM_005254628.3 |
3165 |
Silent Mutation |
CTA,CTG |
L667L |
XP_005254685.1 |
XM_005254633.2 |
3165 |
Silent Mutation |
CTA,CTG |
L657L |
XP_005254690.1 |
XM_006720665.3 |
3165 |
Silent Mutation |
CTA,CTG |
L665L |
XP_006720728.1 |
XM_011521976.2 |
3165 |
Silent Mutation |
CTA,CTG |
L684L |
XP_011520278.1 |
XM_011521977.2 |
3165 |
Silent Mutation |
CTA,CTG |
L683L |
XP_011520279.1 |
XM_011521978.2 |
3165 |
Silent Mutation |
CTA,CTG |
L679L |
XP_011520280.1 |
XM_011521979.2 |
3165 |
Silent Mutation |
CTA,CTG |
L612L |
XP_011520281.1 |
XM_011521980.2 |
3165 |
Silent Mutation |
CTA,CTG |
L673L |
XP_011520282.1 |
XM_011521981.1 |
3165 |
Silent Mutation |
CTA,CTG |
L684L |
XP_011520283.1 |
XM_011521982.2 |
3165 |
Silent Mutation |
CTA,CTG |
L600L |
XP_011520284.1 |
XM_011521983.2 |
3165 |
Silent Mutation |
CTA,CTG |
L595L |
XP_011520285.1 |
XM_017022532.1 |
3165 |
Intron |
|
|
XP_016878021.1 |
View Full Product Details