Product Details

SNP ID
rs139798032
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:75675968 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCTCTCTGGCTTCCCTGCTTCC[A/G]TCCGGGCGGCCTCGGGGACACTGAG
Phenotype
MIM: 601172
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CSPG4 PubMed Links

Gene Details

Gene
CSPG4
Gene Name
chondroitin sulfate proteoglycan 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001897.4 6644 Missense Mutation ACG,ATG T2184M NP_001888.2

View Full Product Details