Product Details

SNP ID
rs141630035
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:75468886 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGCTCAGGGGTCTGGATGCTGAA[C/G]GCCCTCTGGGTCCTCATGCGTGACA
Phenotype
MIM: 600768
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PTPN9 PubMed Links

Gene Details

Gene
PTPN9
Gene Name
protein tyrosine phosphatase, non-receptor type 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002833.3 2180 Silent Mutation GCC,GCG A555A NP_002824.1

View Full Product Details