Product Details

SNP ID
rs142251536
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40767999 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCGTGCGATCAGCTGTTGCCGCT[C/G]GGCCGCCTGGCGCATGCGCATCATG
Phenotype
MIM: 616844 MIM: 602437
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C15orf62 PubMed Links

Gene Details

Gene
C15orf62
Gene Name
chromosome 15 open reading frame 62
There are no transcripts associated with this gene.

Gene
DNAJC17
Gene Name
DnaJ heat shock protein family (Hsp40) member C17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018163.2 888 Missense Mutation CAG,GAG Q286E NP_060633.1
XM_017022401.1 888 Missense Mutation CAG,GAG Q290E XP_016877890.1
XM_017022402.1 888 Intron XP_016877891.1
Gene
GCHFR
Gene Name
GTP cyclohydrolase I feedback regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005258.2 888 Intron NP_005249.1

View Full Product Details