Product Details

SNP ID
rs142561291
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:34103381 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGCTATATGTACTTTGTGAAAGT[C/G]AGTCTGGTTATGTGTGGAATGCGCT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EMC7 PubMed Links

Gene Details

Gene
EMC7
Gene Name
ER membrane protein complex subunit 7
There are no transcripts associated with this gene.

Gene
PGBD4
Gene Name
piggyBac transposable element derived 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152595.4 1309 Missense Mutation CAG,GAG Q284E NP_689808.2

View Full Product Details