Product Details

SNP ID
rs143399953
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:78003308 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCTGAGCTGAGCTAACTCACCTT[C/T]GGTCCTTCATAAAGGAAAGAGTCCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC91450 PubMed Links

Gene Details

Gene
LOC91450
Gene Name
uncharacterized LOC91450
There are no transcripts associated with this gene.

Gene
TBC1D2B
Gene Name
TBC1 domain family member 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015079.5 2571 Silent Mutation CCA,CCG P857P NP_055894.6
NM_144572.1 2571 Silent Mutation CCA,CCG P857P NP_653173.1
XM_011521387.2 2571 Silent Mutation CCA,CCG P820P XP_011519689.1
XM_017022022.1 2571 Silent Mutation CCA,CCG P627P XP_016877511.1

View Full Product Details