Product Details

SNP ID
rs143590623
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40471921 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGAGATCCGAGAGTACCAGCAACG[A/C]TATGGGGCTGAGATAGTGAGGCGGT
Phenotype
MIM: 613880 MIM: 608429
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
BAHD1 PubMed Links

Gene Details

Gene
BAHD1
Gene Name
bromo adjacent homology domain containing 1
There are no transcripts associated with this gene.

Gene
CHST14
Gene Name
carbohydrate sulfotransferase 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_130468.3 961 Silent Mutation CGA,CGC R236R NP_569735.1

View Full Product Details