Product Details

SNP ID
rs144146450
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:75843819 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCAGCCCGCACTCGCTGCCGCC[A/G]CCACTCACGCTCCACTGCAACATCA
Phenotype
MIM: 612501
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UBE2Q2 PubMed Links

Gene Details

Gene
UBE2Q2
Gene Name
ubiquitin conjugating enzyme E2 Q2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145335.1 534 Intron NP_001138807.1
NM_001284382.1 534 Silent Mutation CCA,CCG P51P NP_001271311.1
NM_173469.3 534 Silent Mutation CCA,CCG P51P NP_775740.1
XM_005254787.1 534 Silent Mutation CCA,CCG P51P XP_005254844.1
XM_005254788.1 534 Silent Mutation CCA,CCG P51P XP_005254845.1
XM_006720770.1 534 Silent Mutation CCA,CCG P51P XP_006720833.1
XM_011522228.1 534 Intron XP_011520530.1
XM_017022726.1 534 Intron XP_016878215.1
XM_017022727.1 534 Silent Mutation CCA,CCG P51P XP_016878216.1
XM_017022728.1 534 Silent Mutation CCA,CCG P51P XP_016878217.1

View Full Product Details