Product Details

SNP ID
rs145288136
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:56429134 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCACAAATTTCACTCCTTTGTTG[A/G]TATACTTTCCTGAACTCTTCACCAA
Phenotype
MIM: 610766
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MNS1 PubMed Links

Gene Details

Gene
MNS1
Gene Name
meiosis specific nuclear structural 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018365.2 2063 Silent Mutation TAC,TAT Y485Y NP_060835.1
Gene
TEX9
Gene Name
testis expressed 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286449.1 2063 Intron NP_001273378.1
NM_198524.2 2063 Intron NP_940926.1
XM_005254359.4 2063 Intron XP_005254416.1
XM_005254361.3 2063 UTR 3 XP_005254418.1
XM_011521530.2 2063 Intron XP_011519832.1
XM_017022160.1 2063 UTR 3 XP_016877649.1
XM_017022161.1 2063 Intron XP_016877650.1
XM_017022162.1 2063 UTR 3 XP_016877651.1
XM_017022163.1 2063 UTR 3 XP_016877652.1
XM_017022164.1 2063 UTR 3 XP_016877653.1
XM_017022166.1 2063 Intron XP_016877655.1
XM_017022167.1 2063 UTR 3 XP_016877656.1

View Full Product Details