Product Details

SNP ID
rs147198080
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:42068814 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCCTGGCTGGTCTGCACGTTGTA[C/G]TCACTGAGCCGCAGCAGGCGCTCGA
Phenotype
MIM: 612864
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PLA2G4D PubMed Links

Gene Details

Gene
PLA2G4D
Gene Name
phospholipase A2 group IVD
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178034.3 2453 Missense Mutation GAC,GAG D786E NP_828848.3
XM_011521467.2 2453 Missense Mutation GAC,GAG D730E XP_011519769.1

View Full Product Details