Product Details

SNP ID
rs147428832
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:64963704 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACCATTGATGGGTCAATGGCCGC[A/G]TATTTGGTTCCATGGAATTGCAGCA
Phenotype
MIM: 608181
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKDD1A PubMed Links

Gene Details

Gene
ANKDD1A
Gene Name
ankyrin repeat and death domain containing 1A
There are no transcripts associated with this gene.

Gene
SPG21
Gene Name
spastic paraplegia 21 (autosomal recessive, Mast syndrome)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127889.4 1068 Silent Mutation TAC,TAT Y281Y NP_001121361.1
NM_001127890.4 1068 Silent Mutation TAC,TAT Y254Y NP_001121362.1
NM_016630.6 1068 Silent Mutation TAC,TAT Y281Y NP_057714.1
XM_005254437.4 1068 Silent Mutation TAC,TAT Y281Y XP_005254494.1
XM_017022297.1 1068 Silent Mutation TAC,TAT Y281Y XP_016877786.1
XM_017022298.1 1068 Silent Mutation TAC,TAT Y281Y XP_016877787.1

View Full Product Details