Product Details

SNP ID
rs148498973
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:66336983 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATCTAGCTTCAGTAATATTTCTG[C/G]ATGTAGCATCAAGTTGCTGTTGCAC
Phenotype
MIM: 614183 MIM: 610716
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
DIS3L PubMed Links
Additional Information
For this assay, SNP(s) [rs34848112,rs3759786] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
DIS3L
Gene Name
DIS3 like exosome 3'-5' exoribonuclease
There are no transcripts associated with this gene.

Gene
TIPIN
Gene Name
TIMELESS interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289986.1 1150 Intron NP_001276915.1
NM_017858.2 1150 Intron NP_060328.2
XM_005254521.2 1150 Missense Mutation TCC,TGC S294C XP_005254578.1
XM_005254522.4 1150 Missense Mutation TCC,TGC S294C XP_005254579.1
XM_006720594.3 1150 Missense Mutation TCC,TGC S294C XP_006720657.1
XM_011521743.1 1150 Missense Mutation TCC,TGC S294C XP_011520045.1
XM_017022390.1 1150 Missense Mutation TCC,TGC S294C XP_016877879.1
XM_017022391.1 1150 Intron XP_016877880.1

View Full Product Details