Product Details

SNP ID
rs148510226
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:76336909 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGTGGATATTATTTTTCATTATC[C/G]TTTTCTGGGTGCTATGGGTGATCAT
Phenotype
MIM: 609481
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ISL2 PubMed Links

Gene Details

Gene
ISL2
Gene Name
ISL LIM homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145805.2 186 Missense Mutation CCT,CGT P9R NP_665804.1
XM_017022505.1 186 Missense Mutation CCT,CGT P9R XP_016877994.1

View Full Product Details