Product Details

SNP ID
rs148669324
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40738022 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATGGGGTTTTCTGGCTGGAGAGC[A/C]ATGGCTTTGTCCACATGCTCCTAAG
Phenotype
MIM: 179617 MIM: 611873
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
RAD51 PubMed Links

Gene Details

Gene
RAD51
Gene Name
RAD51 recombinase
There are no transcripts associated with this gene.

Gene
RMDN3
Gene Name
regulator of microtubule dynamics 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304802.1 1430 Missense Mutation ATG,ATT M356I NP_001291731.1
NM_001323894.1 1430 Missense Mutation ATG,ATT M356I NP_001310823.1
NM_001323895.1 1430 Missense Mutation ATG,ATT M227I NP_001310824.1
NM_001323896.1 1430 Missense Mutation ATG,ATT M382I NP_001310825.1
NM_001323897.1 1430 Missense Mutation ATG,ATT M382I NP_001310826.1
NM_018145.2 1430 Missense Mutation ATG,ATT M356I NP_060615.1
XM_011521755.2 1430 Missense Mutation ATG,ATT M150I XP_011520057.1

View Full Product Details