Product Details
- SNP ID
-
rs148736697
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:43603355 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGAGCTCCATCTCTGCAATCTGGG[G/T]TGCAGACCAGGCTGCTGGGAATGTC
- Phenotype
-
MIM: 123290
MIM: 606440
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CKMT1B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2260160,rs28495368] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CKMT1B
- Gene Name
- creatine kinase, mitochondrial 1B
There are no transcripts associated with this gene.
- Gene
- STRC
- Gene Name
- stereocilin
View Full Product Details