Product Details

SNP ID
rs148750800
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:68193307 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGGTGTGGGGTGTAAAGTAGATC[A/G]CTCTTCCTGCTCTCAGCTGATGCTG
Phenotype
MIM: 603566
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CALML4 PubMed Links

Gene Details

Gene
CALML4
Gene Name
calmodulin like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031733.2 1861 UTR 3 NP_001026903.2
NM_001286694.1 1861 UTR 3 NP_001273623.1
NM_001286695.1 1861 UTR 3 NP_001273624.1
NM_033429.2 1861 UTR 3 NP_219501.2
Gene
PIAS1
Gene Name
protein inhibitor of activated STAT 1
There are no transcripts associated with this gene.

View Full Product Details