Product Details

SNP ID
rs149062725
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:42162123 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCAAGACCTTTTCCAGGAAGATG[C/T]GTATGTCATTGATCTGAGTGTTTGC
Phenotype
MIM: 612188
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PLA2G4F PubMed Links

Gene Details

Gene
PLA2G4F
Gene Name
phospholipase A2 group IVF
There are no transcripts associated with this gene.

Gene
VPS39
Gene Name
VPS39, HOPS complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301138.1 2547 Missense Mutation CAC,CGC H801R NP_001288067.1
NM_015289.3 2547 Missense Mutation CAC,CGC H790R NP_056104.2
XM_011521403.2 2547 Missense Mutation CAC,CGC H801R XP_011519705.1
XM_011521404.2 2547 Missense Mutation CAC,CGC H790R XP_011519706.1
XM_017022036.1 2547 Missense Mutation CAC,CGC H801R XP_016877525.1
XM_017022037.1 2547 Missense Mutation CAC,CGC H790R XP_016877526.1
XM_017022038.1 2547 Missense Mutation CAC,CGC H666R XP_016877527.1
XM_017022039.1 2547 Missense Mutation CAC,CGC H433R XP_016877528.1

View Full Product Details