Product Details

SNP ID
rs149419129
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43197334 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATAGTTCAGGGGCTACCACGGTGA[C/T]GCTTTTATAGTTGGTTAGGTTCTGG
Phenotype
MIM: 607089 MIM: 177070
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCNDBP1 PubMed Links

Gene Details

Gene
CCNDBP1
Gene Name
cyclin D1 binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012142.4 2076 Intron NP_036274.3
XM_006720448.1 2076 Intron XP_006720511.1
Gene
EPB42
Gene Name
erythrocyte membrane protein band 4.2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000119.2 2076 Missense Mutation ATC,GTC I712V NP_000110.2
NM_001114134.1 2076 Missense Mutation ATC,GTC I682V NP_001107606.1
XM_005254225.1 2076 Missense Mutation ATC,GTC I647V XP_005254282.1
XM_011521349.2 2076 Missense Mutation ATC,GTC I712V XP_011519651.1
XM_011521350.2 2076 Missense Mutation ATC,GTC I712V XP_011519652.1
XM_011521351.2 2076 Missense Mutation ATC,GTC I712V XP_011519653.1
XM_011521352.2 2076 Missense Mutation ATC,GTC I700V XP_011519654.1
XM_011521353.2 2076 Missense Mutation ATC,GTC I677V XP_011519655.1
XM_011521354.2 2076 Missense Mutation ATC,GTC I527V XP_011519656.1

View Full Product Details