Product Details

SNP ID
rs149593184
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:81313321 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAACCGGGTCATGCTGCGGGGGA[A/G]GAAGGAGTCCACCACGTTCTGTGGG
Phenotype
MIM: 603035 MIM: 607050
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL16 PubMed Links

Gene Details

Gene
IL16
Gene Name
interleukin 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172128.1 1176 Intron NP_001165599.1
NM_004513.5 1176 Intron NP_004504.3
NM_172217.3 1176 Intron NP_757366.2
XM_005254342.3 1176 Intron XP_005254399.1
XM_005254346.4 1176 Intron XP_005254403.1
XM_011521519.2 1176 Intron XP_011519821.1
XM_011521520.2 1176 Intron XP_011519822.1
XM_017022140.1 1176 Intron XP_016877629.1
XM_017022141.1 1176 Intron XP_016877630.1
XM_017022142.1 1176 Intron XP_016877631.1
XM_017022143.1 1176 Intron XP_016877632.1
XM_017022144.1 1176 Intron XP_016877633.1
XM_017022145.1 1176 Intron XP_016877634.1
XM_017022146.1 1176 Intron XP_016877635.1
Gene
STARD5
Gene Name
StAR related lipid transfer domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181900.2 1176 Missense Mutation NP_871629.1
XM_006720703.1 1176 Intron XP_006720766.1
XM_017022639.1 1176 Missense Mutation XP_016878128.1

View Full Product Details