Product Details

SNP ID
rs149680224
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:73740199 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCTCACCGTCCCCCTCCTCATC[A/G]TCACAGCACAGAGCATGGTAGAGCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C15orf59 PubMed Links

Gene Details

Gene
C15orf59
Gene Name
chromosome 15 open reading frame 59
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039614.2 828 Silent Mutation GAC,GAT D200D NP_001034703.1
NM_001303254.1 828 Silent Mutation GAC,GAT D172D NP_001290183.1
XM_005254369.4 828 Silent Mutation GAC,GAT D200D XP_005254426.1

View Full Product Details