Product Details
- SNP ID
-
rs150205920
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:58883755 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGTCCGCTTGTGTCCCGTCCATGG[C/T]GTTCAACCACATGTCGCTCCTCAGG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SLTM
PubMed Links
Gene Details
- Gene
- SLTM
- Gene Name
- SAFB like transcription modulator
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001013843.2 |
2774 |
Missense Mutation |
CAC,CGC |
H938R |
NP_001013865.1 |
NM_024755.3 |
2774 |
Missense Mutation |
CAC,CGC |
H956R |
NP_079031.2 |
XM_006720686.3 |
2774 |
Missense Mutation |
CAC,CGC |
H1009R |
XP_006720749.3 |
XM_006720690.1 |
2774 |
Missense Mutation |
CAC,CGC |
H525R |
XP_006720753.1 |
XM_011522022.1 |
2774 |
Missense Mutation |
CAC,CGC |
H1098R |
XP_011520324.1 |
XM_011522023.1 |
2774 |
Missense Mutation |
CAC,CGC |
H1080R |
XP_011520325.1 |
XM_011522024.1 |
2774 |
Missense Mutation |
CAC,CGC |
H1048R |
XP_011520326.1 |
XM_011522025.1 |
2774 |
Missense Mutation |
CAC,CGC |
H1032R |
XP_011520327.1 |
XM_011522026.2 |
2774 |
Missense Mutation |
CAC,CGC |
H1025R |
XP_011520328.2 |
XM_011522027.1 |
2774 |
Missense Mutation |
CAC,CGC |
H1016R |
XP_011520329.1 |
XM_011522028.1 |
2774 |
Missense Mutation |
CAC,CGC |
H998R |
XP_011520330.1 |
XM_011522029.2 |
2774 |
Missense Mutation |
CAC,CGC |
H910R |
XP_011520331.1 |
XM_011522030.2 |
2774 |
Intron |
|
|
XP_011520332.1 |
XM_011522031.2 |
2774 |
Missense Mutation |
CAC,CGC |
H728R |
XP_011520333.1 |
XM_017022576.1 |
2774 |
Missense Mutation |
CAC,CGC |
H888R |
XP_016878065.1 |
XM_017022577.1 |
2774 |
Missense Mutation |
CAC,CGC |
H1007R |
XP_016878066.1 |
XM_017022578.1 |
2774 |
Missense Mutation |
CAC,CGC |
H991R |
XP_016878067.1 |
XM_017022579.1 |
2774 |
Missense Mutation |
CAC,CGC |
H903R |
XP_016878068.1 |
XM_017022580.1 |
2774 |
Missense Mutation |
CAC,CGC |
H525R |
XP_016878069.1 |
View Full Product Details